Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: LSVSRENLN KDLLTAAEKY PNVKMHFNHR LLKCNPEEGM ITVLGSDKVP KDVTCDLIVG CDGAYSTVRS HLMKKPRFDY SQQYIPHGYM ELTIPPKNGD YAMEPNYLHI WPRNTFMMIA LPNMNKSFTC TLFMPFEEFE KLLTSNDVVD FFQKYFPDAI PLIGEKLLVQ DFFLLPAQPM ISVKCSSFHF KSHCVLLGDA AHAIVPFFGQ GMNAGFEDCL VFDELMDKFS NDLSLCLPVF SRLRIPDDHA ISDLSMYNYI EKNMERFLHA IMPSTFIPLY TMVTFSRIRY HEAVQRWHWQ KR (107-407 aa encoded by BC005297)
Kynurenine 3-monooxygenase (KMO; EC 1.14.13.9) is an NADPH-dependent flavin monooxygenase that catalyzes the hydroxylation of the L-tryptophan metabolite L-kynurenine to form L-3-hydroxykynurenine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hK3OH-2; Kynurenine 3-hydroxylase; Kynurenine 3-monooxygenase; unnamed protein product
Gene Aliases: dJ317G22.1; KMO
UniProt ID: (Human) O15229
Entrez Gene ID: (Human) 8564
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support