Search
Search
Bethyl Laboratories
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes the precursor of a subunit of the mitochondrial ribonuclease P, which is involved in 5' processing of mitochondrial tRNAs. The encoded protein may confer RNA-binding capacity to mitochondrial ribonuclease P and may be essential for transcript processing, RNA modification, translation and mitochondrial respiration.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ20432; HBV pre-S2 trans-regulated protein 2; Mitochondrial ribonuclease P protein 1; Mitochondrial RNase P protein 1; mitochondrial RNase P subunit 1; mRNA methyladenosine-N(1)-methyltransferase; Renal carcinoma antigen NY-REN-49; RNA (guanine-9-) methyltransferase domain containing 1; RNA (guanine-9-)-methyltransferase domain-containing protein 1; transactivated by hepatitis B virus pre-S2 protein; tRNA (adenine(9)-N(1))-methyltransferase; tRNA (guanine(9)-N(1))-methyltransferase; tRNA methyltransferase 10 homolog C; unnamed protein product
Gene Aliases: COXPD30; HNYA; MRPP1; RG9MTD1; TRMT10C
UniProt ID: (Human) Q7L0Y3
Entrez Gene ID: (Human) 54931
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support