Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
There is one major form (long chain) and two minor forms (short chain and medium chain) of MCP-4 produced by differential signal peptide cleavage. Thus, the resulting MW of secreted MCP-4 ranges between ~8-9kD. Western blot analysis of recombinant MCP-4/CCL13 protein detects an ~17kD band, likely corresponding to a dimer of MCP-4.
The M809 MCP-4 antibody (clone 3G4) has successfully been paired as the coating antibody in a sandwich ELISA with detection antibody M810 (biotinylated conjugate of clone 8C12). The M809 MCP-4 antibody (biotinylated conjugate of clone 3G4) has also been used successfully in a sandwich ELISA as a detection antibody when paired with M810 (clone 8C12) as the coating antibody.
Monocyte Chemotactic Protein 4 (MCP-4), also called CCL13, is induced by inflammatory proteins such as IL-1 and TNFalpha. MCP-4 is a ligand for three different G protein coupled receptors, CCR2, CCR3 and CCR5. MCP-4 activates signaling in monocytes, T lymphocytes, eosinophils and basophils and this signaling is associated with the allergic response.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-C motif chemokine 13; CCL 13; CCL13; chemokine (C-C motif) ligand 13; CK-beta-10; H-MCP-4; MCP-4; Monocyte chemoattractant protein 4; Monocyte chemotactic protein 4; NCC-1; new CC chemokine 1; small inducible cytokine subfamily A (Cys-Cys), member 13; Small-inducible cytokine A13
Gene Aliases: CCL13; CKb10; MCP-4; MCP4; NCC-1; NCC1; SCYA13; SCYL1
UniProt ID: (Human) Q99616
Entrez Gene ID: (Human) 6357
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support