Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 5
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Nop1p / Fibrillarin was originally identified as a nucleolar protein of bakers yeast, Saccharomyces cerevisiae (accession P15646). The Nop1p protein is essential for yeast viability and is localized in the nucleoli. The human homologue of Nop1p is fibrillarin (accession P22087) a component of the nucleolar small nuclear ribonucleoprotein (snRNP) particle. The human fibrillarin gene is located on chromosome 19 (19q13. 1).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 34 kDa nucleolar scleroderma antigen; 34-kD nucleolar scleroderma antigen; AT20009p; CG5725-PA; CG5725-PB; CG5725-PC; CG5725-PD; CG5725-PE; CG5725-PF; CG5725-PG; FBL; fbl-PA; fbl-PB; fbl-PC; fbl-PD; fbl-PE; fbl-PF; fbl-PG; Histone-glutamine methyltransferase; LD24618p; LP11571p; Nucleolar protein 1; pantothenate kinase; RNA, U3 small nucleolar interacting protein 1; rRNA 2'-O-methyltransferase fibrillarin; U3 small nucleolar RNA-associated protein NOP1; U6 snRNA 2'-O-methyltransferase fibrillarin; unnamed protein product
Gene Aliases: AL022665; CELE_T01C3.7; CG5725; D2870; Dmel\CG5725; Dmel_CG5725; dPANK; dPANK/Fbl; FBL; FIB; fib-1; FIB1; FLRN; LOT3; NOP1; PANK; RNU3IP1; T01C3.7; YDL014W
UniProt ID: (Human) P22087, (Rat) P22509, (Mouse) Q99L58, (Baker's yeast) P15646, (C. elegans) Q22053
Entrez Gene ID: (Human) 2091, (Rat) 292747, (Mouse) 14113, (Baker's yeast) 851548, (Fruit fly) 46234, (C. elegans) 179999
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support