Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 7
MA3-030 detects brain nitric oxide synthase (bNOS), inducible NOS (iNOS) and epithelial NOS (eNOS) in bovine, human, mouse and rat tissues.
MA3-030 has been successfully used in Western blot, immunofluorescence, immunoprecipitation, and immunohistochemical experiments. By Western blot, this antibody detects an ~130 kDa band representing iNOS in samples first induced with interferon (IFN) and lipopolysaccarides (LPS), an ~155 kDa band in tissues expressing bNOS and an ~140 kDa band in tissues expressing eNOS. Immunohistochemical staining of NOS with MA3-030 yields a pattern consistent with that seen in the literature and depends on the tissue being studied and the localization of the isoforms present.
The MA3-030 antigen is purified bovine bNOS.
NOS oxidizes a guanidine nitrogen of arginine releasing nitric oxide in the form of a free radical and citrulline. Nitric oxide thus generated acts as a messenger in diverse functions including vasodilation neurotransmission, anti-tumor and anti-pathogenic activities. NOS is classified under three types: neuronal NOS (nNOS) or brain NOS (bNOS); induciblr NOS (iNOS) or macrophage NOS (mNOS); and endothelial NOS (eNOS). This antibody reacts with iNOS, bNOS and eNOS.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bNOS; calcium-calmodulin independent nitric oxide synthase iNOS; cNOS; Constitutive NOS; EC-NOS; endothelial nitric oxide synthase; endothelial nitric oxide synthase 3; Endothelial NOS; eNOS; HEP-NOS; Hepatocyte NOS; inducible nitric oxide synthase; Inducible NO synthase; Inducible NOS; MAC-NOS; Macrophage NOS; N-NOS; NC-NOS; Neuronal NOS; nitric oxidase synthase; nitric oxide synthase; Nitric oxide synthase 1; nitric oxide synthase 1 (neuronal); nitric oxide synthase 2, inducible; nitric oxide synthase 2, inducible, macrophage; nitric oxide synthase 2A (inducible, hepatocytes); Nitric oxide synthase 3; nitric oxide synthase 3 (endothelial cell); Nitric oxide synthase, brain; Nitric oxide synthase, endothelial; Nitric oxide synthase, inducible; nitric oxide synthase, macrophage; nNOS; NOS type I; NOS type II; NOS type III; NOS, type II; NOSII; NOSIII; Peptidyl-cysteine S-nitrosylase NOS1; Peptidyl-cysteine S-nitrosylase NOS2; unnamed protein product
Gene Aliases: 2310005C01Rik; 2310065A03Rik; Bnos; ECNOS; eNOS; HEP-NOS; i-NOS; IHPS1; INOS; Inosl; N-NOS; NC-NOS; nNOS; NO; NOS; Nos-1; Nos-2; Nos-3; NOS-I; NOS-II; NOS1; NOS2; NOS2A; NOS3
UniProt ID: (Human) P29475, (Human) P35228, (Bovine) Q27995, (Human) P29474, (Rat) P29476, (Mouse) Q9Z0J4, (Mouse) P29477, (Rat) Q06518, (Mouse) P70313, (Rat) Q62600
Entrez Gene ID: (Human) 4842, (Human) 4843, (Bovine) 282876, (Bovine) 287024, (Human) 4846, (Rat) 24598, (Mouse) 18125, (Mouse) 18126, (Rat) 24599, (Mouse) 18127, (Rat) 24600
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support