Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody recognizes the C-terminal region of nesprin-1.
This target has a predicted molecular weight of 1011 kDa.
There are two genes encoding members of a new family of type II integral membrane proteins. Both are ubiquitously expressed, and tissue-specific alternative mRNA initiation and splicing generate at least two major isoforms of each protein, with the smaller isoforms being truncated at the N-terminus. These proteins are called nesprin-l and -2 for nuclear _envelope _spectrin repeat, as they are characterized by the presence of multiple, clustered spectrin repeats, bipartite nuclear localization sequences and a conserved C-terminal, single transmembrane domain. Transient transfection of EGFP-fusion expression constructs demonstrated their localization to the nuclear membrane with a novel C-terminal, TM-domain-containing sequence essential for perinuclear localization. Nesprin-l is developmentally regulated in both smooth and skeletal muscle and is relocalized from the nuclear envelope to the nucleus and cytoplasm during C2Cl2 myoblast differentiation. Nesprins may function as 'dystrophins of the nucleus' to maintain nuclear organization and structura integrity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: actin-binding protein; Syne-1; CPG2 full length; cytoskeletal protein; DKFZp781J13156; Enaptin; enaptin165; FLJ30878; FLJ41140; KASH domain-containing protein 1; KASH1; Myne-1; MYNE1; Myocyte nuclear envelope protein 1; nesprin 1; Nesprin-1; Nuclear envelope spectrin repeat protein 1; RP1-130E4.2; spectrin repeat containing, nuclear envelope 1; synaptic nuclear envelope 1; Synaptic nuclear envelope protein 1; synaptic nuclei expressed gene 1; SYNE 1; Syne-1; syne-1 fragment; unnamed protein product
Gene Aliases: 8B; A330049M09Rik; AMC3; AMCM; ARCA1; BE692247; C130039F11Rik; C6orf98; CPG2; dJ45H2.2; EDMD4; KASH1; KIAA0796; KIAA1262; KIAA1756; mKIAA1756; Myne-1; MYNE1; Nesp1; SCAR8; SYNE1
UniProt ID: (Human) Q8NF91, (Mouse) Q6ZWR6
Entrez Gene ID: (Human) 23345, (Mouse) 64009, (Rat) 499010
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support