Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Reconstitute at 0.5 mg/mL in sterile PBS.
CORIN is a member of the type II transmembrane serine protease class of the trypsin superfamily. Members of this family are composed of multiple structurally distinct domains. CORIN converts pro-atrial natriuretic peptide to biologically active atrial natriuretic peptide, a cardiac hormone that regulates blood volume and pressure. This protein may also function as a pro-brain-type natriuretic peptide convertase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Atrial natriuretic peptide-converting enzyme; Corin; EC 3.4.21; Heart-specific serine proteinase ATC2; HTSP; MGC119742; mosiac serine protease; pro-ANP-convertase; Pro-ANP-converting enzyme; PRSC; serine peptidase; serine protease; Transmembrane protease serine 10; unnamed protein product
Gene Aliases: ATC2; CMH30; CORIN; CRN; Lrp4; PEE5; TMPRSS10
UniProt ID: (Human) Q9Y5Q5
Entrez Gene ID: (Human) 10699
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support