Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: RENSSGARSPR VPADLARSIL ISLPFPPDSL AHRPPSSAPT ESQGLFTVAA PAPGAPSPPA TLAHLLPAPA MYSLLETELK NPVGTPTQAA GTGGPAAPGG AGKSSA
Binds to an epitope located within the peptide sequence LISLPFPPDS as determined by overlapping synthetic peptides.
Highest antigen sequence identity to the following orthologs - mouse 82%, rat 31%.
This gene encodes a member of the SOX family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. Mutations in this gene have been associated with X-linked mental retardation with growth hormone deficiency.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: soxB; SRY (sex determining region Y)-box 3; SRY-box 3; Transcription factor SOX-3
Gene Aliases: GHDX; MRGH; PHP; PHPX; SOX3; SOXB
UniProt ID: (Human) P41225
Entrez Gene ID: (Human) 6658
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support