Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Positive Samples: HeLa, HT-29, HepG2
Immunogen sequence: AGLGPVAEVK SSPVGGGPRG PAGPALGRHW GAGWDGGHHY PEMDARQERV EVLPQARASW ESLDEEWRAP QAGSRTPVPS TVQANTFEFA DAEEDDEVKV
CDC42 is a member of the Rho GTPase family that regulates multiple cellular activities, including actin polymerization. The protein encoded by this gene is a CDC42 binding protein that mediates actin cytoskeleton reorganization at the plasma membrane. This protein is secreted and is primarily found in bone marrow.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 55 kDa bone marrow stromal/endothelial cell protein; binder of Rho GTPase 5; Binder of Rho GTPases 5; CDC42 effector protein (Rho GTPase binding) 1; Cdc42 effector protein 1; cdc42ep1 {ECO:0000312|EMBL:AAI28745.1}; MSE55; serum constituent protein; Serum protein MSE55; unnamed protein product
Gene Aliases: 1810058K22Rik; AA980734; BORG5; CDC42EP1; CEP1; MSE55
UniProt ID: (Human) Q00587, (Rat) A1A5P0, (Mouse) Q91W92
Entrez Gene ID: (Human) 11135, (Rat) 315121, (Mouse) 104445
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support