Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
1 µg/mL of MA5-45435 was sufficient for detection of Slo2.2 in 10 µg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 140kDa. Weak human detection.approximately Does not cross-react with KCNT2/Slo2.1/Slick.
Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a sodium-activated potassium channel subunit which is thought to function in ion conductance and developmental signaling pathways. Mutations in this gene cause the early-onset epileptic disorders, malignant migrating partial seizures of infancy and autosomal dominant nocturnal frontal lobe epilepsy. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KCa4.1; KCa4.1KIAA1422FLJ41282; KNa1.1; Potassium channel subfamily T member 1; potassium channel subunit (Slack); potassium channel, sodium activated subfamily T, member 1; potassium channel, subfamily T, member 1; Sequence like a calcium-activated K+ channel; Sequence like a calcium-activated potassium channel subunit; Slo2.2; Sodium and chloride-activated ATP-sensitive potassium channel Slo2.2; unnamed protein product
Gene Aliases: bA100C15.2; C030030G16Rik; DEE14; EIEE14; ENFL5; KCa4.1; KCNT1; KIAA1422; KNa1.1; rSlo2; SLACK; slo2; Slo2.2
UniProt ID: (Human) Q5JUK3, (Mouse) Q6ZPR4, (Rat) Q9Z258
Entrez Gene ID: (Human) 57582, (Mouse) 227632, (Rat) 60444
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support