Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: IDEYCVQQLK EFEGKTLVSV TKEGLELPED EEEKKKQEEK KTKFENLCKI MKDILEKKVE KVVVSNRLVT SPCCIVTSTY GWTANMERIM KAQALRDNST MGYMAAKKHL EINPDHSIIE TLRQKAEADK NDKSVKDLVI LLYETALLSS GFSLEDPQTH ANRIYRMIKL GLGIDEDDPT ADDTSAAVTE EMPPLEGDDD
Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%, Pig - 100%, Cynomolgus monkey - 100%.
HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. There are 2 major cytosolic HSP90 proteins, HSP90AA1, an inducible form, and HSP90AB1 and mitochondria (Chen et al., 2005 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 90kDa heat shock protein; epididymis luminal secretory protein 52; epididymis secretory sperm binding protein Li 65p; Heat shock 84 kDa; Heat shock 86 kDa; heat shock 90kD protein 1, alpha; heat shock 90kD protein 1, alpha-like 4; heat shock 90kD protein 1, beta; heat shock 90kD protein, alpha-like 4; heat shock 90kDa protein 1, alpha; heat shock 90kDa protein 1, beta; Heat shock protein; heat shock protein 1, alpha; heat shock protein 1, beta; heat shock protein 86; heat shock protein 90 kDa; heat shock protein 90kDa alpha (cytosolic), class A member 1; heat shock protein 90kDa alpha (cytosolic), class B member 1; heat shock protein 90kDa alpha family class A member 1; heat shock protein 90kDa alpha family class B member 1; Heat shock protein family C member 1; Heat shock protein family C member 3; Heat shock protein HSP 90-alpha; Heat shock protein HSP 90-beta; heat shock protein, 1; heat shock protein, 84 kDa 1; heat shock protein, 86 kDa 1; heat shock protein, 89 kDa; HSP; HSP 84; HSP 86; HSP 90; HSP84; HSP86; HSP90-beta; LAP-2; Lipopolysaccharide-associated protein 2; LPS-associated protein 2; Renal carcinoma antigen NY-REN-38; retinal degeneration slow protein; TSTA; Tumor-specific transplantation 84 kDa antigen; Tumor-specific transplantation 86 kDa antigen; unnamed protein product
Gene Aliases: 86kDa; 89kDa; 90kDa; AL022974; AL024080; AL024147; C81438; D6S182; EL52; HEL-S-65p; Hsp103; hsp4; HSP84; Hsp84-1; HSP86; Hsp86-1; Hsp89; HSP89A; Hsp90; HSP90-BETA; HSP90A; HSP90AA1; HSP90AB1; HSP90B; HSP90N; HSPC1; HSPC2; HSPC3; HSPCA; HSPCAL1; HSPCAL4; HSPCB; HSPN; LAP-2; LAP2
UniProt ID: (Human) P07900, (Human) P08238, (Mouse) P07901, (Rat) P82995, (Mouse) P11499, (Rat) P34058
Entrez Gene ID: (Human) 3320, (Human) 3326, (Mouse) 15519, (Rat) 299331, (Mouse) 15516, (Rat) 301252
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support