Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Antibody detects endogenous levels of total MED27.
The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by this gene is a subunit of the CRSP complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e.g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cofactor required for Sp1 transcriptional activation subunit 8; cofactor required for Sp1 transcriptional activation, subunit 8; cofactor required for Sp1 transcriptional activation, subunit 8, 34kDa; CRSP complex subunit 8; CRSP8; epididymis secretory sperm binding protein; Mediator complex subunit 27; Mediator of RNA polymerase II transcription subunit 27; MGC11274; P37 TRAP/SMCC/PC2 subunit; transcriptional co-activator CRSP8 homolog; Transcriptional coactivator CRSP34; unnamed protein product
Gene Aliases: 1500015J03Rik; 2310042P07Rik; AA682045; CRAP34; CRSP34; CRSP8; D2Ertd434e; MED27; MED3; NEDSCAC; TRAP37
UniProt ID: (Human) Q6P2C8, (Mouse) Q9DB40
Entrez Gene ID: (Human) 9442, (Mouse) 68975
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support