Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 5
Antibody detects endogenous levels of total TMX2.
This gene encodes a member of the disulfide isomerase (PDI) family of endoplasmic reticulum (ER) proteins that catalyze protein folding and thiol-disulfide interchange reactions. The encoded protein has an N-terminal ER-signal sequence, a catalytically active thioredoxin domain, one transmembrane domain and a C-terminal ER-retention sequence. This protein is enriched on the mitochondria-associated-membrane of the ER via palmitoylation of two of its cytosolically exposed cysteines.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell proliferation-inducing gene 26 protein; growth-inhibiting gene 11; PIG26; protein disulfide isomerase family A, member 12; thioredoxin domain containing 14; Thioredoxin domain-containing protein 14; Thioredoxin-related transmembrane protein 2; unnamed protein product
Gene Aliases: 2310042M24Rik; AA589631; CGI-31; My009; NEDMCMS; PDIA12; PIG26; PSEC0045; TMX2; TXNDC14; UNQ237/PRO270
UniProt ID: (Human) Q9Y320, (Mouse) Q9D710
Entrez Gene ID: (Human) 51075, (Mouse) 66958
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support