Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Antibody detects endogenous levels of total PPP1R1C.
Protein phosphatase-1.[supplied by OMIM, Feb 2010].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Inhibitor-5 of protein phosphatase 1; IPP5; PP1 inhibitor protein 5; PP1I; protein phosphatase 1 regulatory subunit 1A; Protein phosphatase 1 regulatory subunit 1C
Gene Aliases: 4930565M23Rik; 4930579P15Rik; IPP5; PPP1R1C; RGD1562095
UniProt ID: (Human) Q8WVI7, (Mouse) Q8BKK4
Entrez Gene ID: (Human) 151242, (Mouse) 75276, (Rat) 499818
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support