Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Antibody detects endogenous levels of MuSK only when phosphorylated at Tyr751.
MUSK plays an important and selective role in many aspects of skeletal muscle development and function through the initial step of postsynaptic differentiation and formation of end-plate bands in cooperation with Rapsyn. Communication between cells is often mediated by receptors on the surface of the target cell that are triggered by specific ligands released by other cells. Receptor tyrosine kinase activity is strictly regulated by the binding of the appropriate ligand to the extracellular active site of the receptor. MuSK has been shown to be part of the agrin signaling complex and plays an important role in the development and architectural maintenance of the neuromuscular junction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC126323; MGC126324; muscle localized kinase 2; muscle specific kinase (neural fold/somite kinase 1); Muscle, skeletal receptor tyrosine protein kinase; Muscle, skeletal receptor tyrosine-protein kinase; muscle, skeletal, receptor tyrosine kinase; muscle-specific kinase receptor; muscle-specific protein kinase secretory isoform; muscle-specific tyrosine kinase receptor MuSK; Muscle-specific tyrosine protein kinase receptor; Muscle-specific tyrosine-protein kinase receptor; MuSK; RP23-170A3.1
Gene Aliases: CMS9; FADS; FADS1; Mdk4; Mlk; MUSK; Nsk1; Nsk2; Nsk3
UniProt ID: (Human) O15146, (Mouse) Q61006, (Rat) Q62838
Entrez Gene ID: (Human) 4593, (Mouse) 18198, (Rat) 81725
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support