Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 14
Antibody detects endogenous levels of total TAF12.
TAF12 is involved in the control of transcription by RNA polymerase II involves the basal transcription machinery which is a collection of proteins. These proteins with RNA polymerase II, assemble into complexes which are modulated by transactivator proteins that bind to cis-regulatory elements located adjacent to the transcription start site. Some modulators interact directly with the basal complex, whereas others may act as bridging proteins linking transactivators to the basal transcription factors. Some of these associated factors are weakly attached while others are tightly associated with TBP in the TFIID complex. Among the latter are the TAF proteins. Different TAFs are predicted to mediate the function of distinct transcriptional activators for a variety of gene promoters and RNA polymerases. TAF12 interacts directly with TBP as well as with TAF2I.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 20 kDa; TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 20kDa; TAFII-20; TAFII-20/TAFII-15; TAFII20; TAFII20/TAFII15; TATA box binding protein (TBP)-associated factor, RNA polymerase II, J, 20kD; Transcription initiation factor TFIID 20 kDa subunits; Transcription initiation factor TFIID 20/15 kDa subunits; Transcription initiation factor TFIID subunit 12
Gene Aliases: 20kDa; 2810422D08Rik; AW557038; TAF12; TAF15; TAF2J; TAFII20
UniProt ID: (Human) Q16514, (Mouse) Q8VE65
Entrez Gene ID: (Human) 6883, (Mouse) 66464, (Rat) 682902
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support