Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Involved in regulating cell migration through association with the actin cytoskeleton. Has an essential role in the maintenance of Z line and sarcomere integrity.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: autoantigen; F-actin binding protein; F-actin-binding protein; Nelin; Nexilin; nexilin alpha; nexilin beta; nexilin gamma; nexn; nexn {ECO:0000312|RGD:708354}; nF actin binding protein; unnamed protein product
Gene Aliases: 1110046H09Rik; AA553326; CMH20; NELIN; NEXN
UniProt ID: (Human) Q0ZGT2, (Rat) Q9Z2J4, (Mouse) Q7TPW1
Entrez Gene ID: (Human) 91624, (Rat) 246172, (Mouse) 68810
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support