Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: ILNELKPEQV EQLKLIMSKR YDGSQQALDL KGLRSDPDLV AQNIDVVLNR RSCMAATLRI IEENIPELLS LNLSNNRLYR LDDMSSIVQK APNLKILNLS G
The constitutive transport element (CTE) of type D retroviruses serves as a signal of nuclear export for unspliced viral RNAs. TAP (also known as NXF1) mediates the export of CTE-containing simian type D retroviral RNAs through binding directly to the CTE. TAP is associated with a recognized mRNA export pathway and is a member of the multigene family of NXF proteins. NXF proteins belong to an evolutionarily conserved family of proteins, which are characterized by a leucine-rich-repeat domain (LRR) followed by a region known as the Nuclear Transport Factor 2 (NTF2)-like domain.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CTE RNA export factor; Mex 67 homolog; modifier of vibrator 1; mRNA export factor TAP; Nuclear RNA export factor 1; nuclear RNA export factor 1 homolog; TAP; tip associated protein; tip associating protein; Tip-associated protein; Tip-associating protein; unnamed protein product; Vdp
Gene Aliases: MEX67; Mex67h; Mvb1; NXF1; TAP
UniProt ID: (Human) Q9UBU9, (Rat) O88984, (Mouse) Q99JX7
Entrez Gene ID: (Human) 10482, (Rat) 59087, (Mouse) 53319
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support