Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: RLSYS
PRKCD (Protein Kinase C Delta) is a Protein Coding gene. Diseases associated with PRKCD includeAutoimmune Lymphoproliferative Syndrome, Type IiiandSystemic Lupus Erythematosus 16. Among its related pathways areRole of Calcineurin dependent NFAT signaling in lymphocytesandWnt Signaling Pathway and Pluripotency. Gene Ontology (GO) annotations related to this gene includetransferase activity, transferring phosphorus containing groupsandprotein tyrosine kinase activity. An important paralog of this gene isPRKCQ (PKC theta).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: nPKC-delta; nPKC-theta; PKC theta; PKCdeltaVIII; Protein kinase C delta type; protein kinase C delta variant IX; protein kinase C delta VIII; Protein kinase C theta type; protein kinase C, delta IV; protein kinase C, delta V; protein kinase C[d]; SDK-1; Tyrosine-protein kinase PRKCD; unnamed protein product
Gene Aliases: A130035A12Rik; AI385711; ALPS3; AW494342; CVID9; D14Ertd420e; MAY1; nPKC-delta; nPKC-theta; PKC-0; PKC-theta; PKC[d]; PKCD; PKCdelta; Pkcq; PKCtheta; PRKCD; PRKCQ; PRKCT
UniProt ID: (Human) Q05655, (Human) Q04759, (Mouse) P28867, (Mouse) Q02111
Entrez Gene ID: (Human) 5580, (Human) 5588, (Mouse) 18753, (Mouse) 18761
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support