Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sulfate groups in carbohydrates confer highly specific functions on glycoproteins, glycolipids, and proteoglycans and are critical for cell-cell interaction, signal transduction, and embryonic development. Sulfotransferases, such as CHST9, carry out sulfation of carbohydrates (Hiraoka et al., 2001 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 9; carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 9 isoform CHST9_1; carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 9 isoform CHST9_2; Carbohydrate sulfotransferase 9; GalNAc-4-O-sulfotransferase; GalNAc-4-O-sulfotransferase (Met86); GalNAc-4-O-sulfotransferase 2; GalNAc-4-ST2; GalNAc-4-sulfotransferase 2; GalNAc4ST-2; N-acetylgalactosamine 4-O-sulfotransferase 2; N-acetylgalactosamine-4-O-sulfotransferase 2; unnamed protein product
Gene Aliases: 5430438D01Rik; CHST9; GALNAC4ST-2; GalNAc4ST2; UNQ2549/PRO6175
UniProt ID: (Human) Q7L1S5, (Mouse) Q76EC5
Entrez Gene ID: (Human) 83539, (Mouse) 71367
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support