Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MRN complex-interacting protein; MRN-interacting protein; unnamed protein product; UPF0544 protein C5orf45; UPF0544 protein C5orf45 homolog
Gene Aliases: 2900016G09Rik; AI851514; AW475975; C5orf45; MRNIP
UniProt ID: (Mouse) Q9D1F5, (Human) Q6NTE8
Entrez Gene ID: (Mouse) 68067, (Human) 51149
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support