Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
MTCH1 is a potential mitochondrial transporter. It may play a role in apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cell proliferation-inducing protein 60; Mitochondrial carrier homolog 1; MTCH1a; Presenilin-associated protein; PSAP; PS-1 associated protein; PDZ-like protein; solute carrier family 25, member 49; unnamed protein product
Gene Aliases: CGI-64; MTCH1; PIG60; PSAP; SLC25A49; UNQ1871/PRO4314
UniProt ID: (Human) Q9NZJ7
Entrez Gene ID: (Human) 23787
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support