Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Antibody detects endogenous levels of total CNGA2.
CNGA2 represents the alpha subunit of a cyclic nucleotide-gated olfactory channel. The encoded protein contains a carboxy-terminal leucine zipper that mediates channel formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CNCalpha3; cng channel; CNG channel 2; CNG channel alpha-2; CNG-2; CNG2; cyclic nucleotide gated channel 4; cyclic nucleotide gated channel alpha 2; cyclic nucleotide-gated cation channel 2; Cyclic nucleotide-gated channel alpha 3; Cyclic nucleotide-gated channel alpha-2; cyclic nucleotide-gated olfactory channel; cyclic nucleotide-gated olfactory channel subunit OCNC1; cyclic-nucleotide-gated cation channel 2; OCNC1; Olfactory cyclic nucleotide-gated channel subunit 1; unnamed protein product
Gene Aliases: CNCA; CNCA1; CNCG2; Cncg4; CNG2; CNGA2; OCNC1; OCNCa; OCNCALPHA; OLFCH
UniProt ID: (Human) Q16280, (Rat) Q00195, (Mouse) Q62398
Entrez Gene ID: (Human) 1260, (Rat) 25411, (Mouse) 12789
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support