Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 6
Antibody detects endogenous levels of total KCNK9.
KCNK9 is one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. This open channel is highly expressed in the cerebellum. It is inhibited by extracellular acidification and arachidonic acid, and strongly inhibited by phorbol 12-myristate 13-acetate.This gene encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. This open channel is highly expressed in the cerebellum. It is inhibited by extracellular acidification and arachidonic acid, and strongly inhibited by phorbol 12-myristate 13-acetate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2P; Acid-sensitive potassium channel protein TASK-3; leak channel; MGC138268; MGC138270; potassium 2-pore domain leak channel TASK3; Potassium channel subfamily K member 9; potassium channel subfamily K member 9 (Task-3); potassium channel, subfamily K, member 9 (Task-3); potassium channel, two pore domain subfamily K, member 9; TWIK-related acid-sensitive K(+) channel 3; TWIK-related acid-sensitive K+ 3; Two pore K(+) channel KT3.2; Two pore potassium channel KT3.2
Gene Aliases: BIBARS; K2p9.1; KCNK9; KT3.2; TASK-3; TASK3; TASK32
UniProt ID: (Human) Q9NPC2, (Rat) Q9ES08, (Mouse) Q3LS21
Entrez Gene ID: (Human) 51305, (Rat) 84429, (Mouse) 223604
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support