Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Positive Samples: Rat lung
Immunogen sequence: DIVVEVDHGK TFSCHRNVLA AISPYFRSMF TSGLTESTQK EVRIVGVEAE SMDLVLNYAY TSRVILTEAN VQALFTAASI FQIPSIQDQC AKYMISHLDP QNSIGVFIFA DHYGHQELGD RSKEYIRKKF LCVTKEQEFL QLTKDQLISI LDSDDLNVDR EEHVYESIIR WFEHEQNERE VHLPEIFAKC IRFPLMEDTF IEKIPPQFAQ AIAKSCVEKG PSNTNGCTQR L
KBTBD8 (Kelch Repeat And BTB Domain Containing 8) is a Protein Coding gene. Diseases associated with KBTBD8 include Treacher Collins Syndrome 1. Among its related pathways are Class I MHC mediated antigen processing and presentation and Innate Immune System. An important paralog of this gene is KBTBD2.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kbtbd8 {ECO:0000312|EMBL:AAI60902.1, ECO:0000312|RGD:1563166}; Kelch repeat and BTB domain-containing protein 8; kelch repeat and BTB domain-containing protein 8 {ECO:0000250|UniProtKB:Q8NFY9}; SSEC 51; T-cell activation kelch repeat protein
Gene Aliases: AU040152; Kbtbd8; Kiaa1842; mKIAA1842; RGD1563166; SSEC-51; SSEC51; Takrp
UniProt ID: (Rat) B1H285, (Mouse) Q3UQV5
Entrez Gene ID: (Rat) 500262, (Mouse) 243574
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support