Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Positive Samples: HeLa, 22Rv1, Mouse uterus, Mouse spleen
Immunogen sequence: FAVGCPLLLL WPFLCESQGL RKRQQPPGNE ADARVRPEEE EEPLMEMRLR DAPQHFYAAL LQLGLKYLFI LGIQILACAL AASILRRHLM VWKVFAPKFI F
This gene encodes a protein that is involved in glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI-anchor is a glycolipid which contains three mannose molecules in its core backbone. The GPI-anchor is found on many blood cells and serves to anchor proteins to the cell surface. This protein is involved in the transfer of ethanolaminephosphate (EtNP) to the third mannose in GPI. At least two alternatively spliced transcripts encoding distinct isoforms have been found for this gene.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GPI ethanolamine phosphate transferase 3, catalytic subunit; Phosphatidylinositol-glycan biosynthesis class O protein; PIG-O; RP11-182N22.4
Gene Aliases: hGPCR43; HPMRS2; PIG-O; PIGO; UNQ632/PRO1249
UniProt ID: (Human) Q8TEQ8
Entrez Gene ID: (Human) 84720, (Mouse) 56703
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support