Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Positive Samples: HL-60, A-549, 22Rv1, SW480, Mouse ovary
Immunogen sequence: MATIEEIAHQ IIEQQMGEIV TEQQTGQKIQ IVTALDHNTQ GKQFILTNHD GSTPSKVILA RQDSTPGKVF LTTPDAAGVN QLFFTTPDLS AQHLQLLTDN SPDQGPNKVF DLCVVCGDKA SGRHYGAVTC EGCKGFFKRS IRKNLVYSCR GSKDCIINKH HRNRCQYCRL QRCIAFGMKQ DSVQCERKPI EVSREKSSNC AASTEKIYIR KDLRSPLTAT PTFVTDSEST RSTGLLDSGM
This gene encodes a nuclear hormone receptor characterized by a highly conserved DNA binding domain (DBD), a variable hinge region, and a carboxy-terminal ligand binding domain (LBD) that is typical for all members of the steroid/thyroid hormone receptor superfamily. This protein also belongs to a large family of ligand-inducible transcription factors that regulate gene expression by binding to specific DNA sequences within promoters of target genes. Multiple alternatively spliced transcript variants have been described, but the full-length nature of some of these variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 80-3 cNDA; early embryonic nuclear receptor; mTR2; nr2c1; NR2C1v1; NR2C1v2; Nuclear receptor subfamily 2 group C member 1; nuclear receptor subfamily 2, group H, member 1; Orphan nuclear receptor TR2; orphan receptor, TR2-11; steroid receptor TR2; steroid receptor TR2-11; steroid receptor TR2-9; Testicular receptor 2; TR2 nuclear hormone receptor; unnamed protein product
Gene Aliases: 4831444H07Rik; 80.3; Eenr; NR2C1; TR2; Tr2-11
UniProt ID: (Human) P13056, (Mouse) Q505F1
Entrez Gene ID: (Human) 7181, (Mouse) 22025
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support