Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Positive test controls include: HeLa, K-562, NIH/3T3, Mouse liver, Mouse heart, Rat liver. The target is usually found in the following locations: Cell membrane, Early endosome membrane, Late endosome membrane, Single-pass type I membrane protein.
Immunogen sequence: MLRGGRRGQL GWHSWAAGPG SLLAWLILAS AGAAPCPDAC CPHGSSGLRC TRDGALDSLH HLPGAENLTE LYIENQQHLQ HLELRDLRGL GELRNLTIVK SGLRFVAPDA FHFTPRLSRL NLSFNALESL SWKTVQGLSL QELVLSGNPL HCSCALRWLQ RWEEEGLGGV PEQKLQCHGQ GPLAHMPNAS CGVPTLKVQV PNASVDVGDD VLLRCQVEGR GLEQAGWILT ELEQSATVMK SGGLPSLGLT LANVTSDLNR KNVTCWAEND VGRAEVSVQV
NTRK1 (TRKA) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system. TrkA is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. The presence of TrkA leads to cell differentiation and may play a role in specifying sensory neuron subtypes. Mutations in TrkA have been associated with congenital insensitivity to pain, anhidrosis, self-mutilating behavior, mental retardation and cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp781I14186; EC 2.7.10.1; gp140trk; High affinity nerve growth factor receptor; kinase TrkA; Neurotrophic tyrosine kinase receptor type 1; neurotrophic tyrosine kinase, receptor, type 1; Oncogene TRK; p140-TrkA; Slow nerve growth factor receptor; trk; trk tyrosine-specific protein kinase; Trk-A; TRK1-transforming tyrosine kinase protein; TrkA neurotrophin receptor; trkA proto-oncogene receptor; tropomyosin receptor kinase A; Tropomyosin-related kinase A; Tyrosine kinase receptor; Tyrosine kinase receptor A; unnamed protein product
Gene Aliases: C80751; MTC; NTRK1; p140-TrkA; Tkr; TRK; Trk-A; TRK1; TRKA
UniProt ID: (Human) P04629, (Mouse) Q3UFB7, (Rat) P35739
Entrez Gene ID: (Human) 4914, (Mouse) 18211, (Rat) 59109
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support