Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Positive test controls include: U-87MG, LO2, HeLa, Jurkat. The target is usually found in the following locations: Basolateral cell membrane, Cell junction, Cell membrane, Endoplasmic reticulum membrane, Membrane, Peripheral membrane protein, postsynaptic cell membrane, postsynaptic density, sarcolemma, synapse.
Immunogen sequence: MPVRKQDTQR ALHLLEEYRS KLSQTEDRQL RSSIERVINI FQSNLFQALI DIQEFYEVTL LDNPKCIDRS KPSEPIQPVN TWEISSLPSS TVTSETLPSS LSPSVEKYRY QDEDTPPQEH ISPQITNEVI GPELVHVSEK NLSEIENVHG FVSHSHISPI KPTEAVLPSP PTVPVIPVLP VPAENTVILP
This gene encodes a multi-domain scaffolding protein that is required for normal development. This protein may have a role in septate junction formation, signal transduction, cell proliferation, synaptogenesis and lymphocyte activation. Several alternatively spliced transcript variants encoding different isoforms have been described for this gene, but the full-length nature of some of the variants is not known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: discs large homolog 1, scribble cell polarity complex component; Disks large homolog 1; dJ1061C18.1.1; DKFZp761P0818; DKFZp781B0426; Drosophila discs-large tumor suppressor homologue (synapse associated protein); E-dlg/SAP97; Embryo-dlg/synapse-associated protein 97; hDlg; presynaptic protein SAP97; SAP-97; Synapse-associated protein 97; unnamed protein product
Gene Aliases: B130052P05Rik; DLG1; DLGH1; E-dlg/SAP97; hdlg; mKIAA4187; SAP-97; SAP97
UniProt ID: (Human) Q12959, (Rat) Q62696, (Mouse) Q811D0
Entrez Gene ID: (Human) 1739, (Rat) 25252, (Mouse) 13383
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support