Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Recommended positive controls: A549, HeLa, HepG2, HCT116, mouse brain.
Predicted reactivity: Mouse (89%), Rat (92%), Bovine (94%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Ubiquitin-specific protease that specifically deubiquitinates monoubiquitinated DNA polymerase beta (POLB), stabilizing POLB thereby playing a role in base-excision repair (BER). Acts as a regulator of cell growth and genome integrity. May also indirectly regulate CDC25A expression at a transcriptional level. [UniProt]
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme 47; Trf (TATA binding protein-related factor)-proximal homolog; Ubiquitin carboxyl-terminal hydrolase 47; ubiquitin specific protease 47; Ubiquitin thioesterase 47; ubiquitin thiolesterase 47; Ubiquitin-specific-processing protease 47; unnamed protein product
Gene Aliases: 4930502N04Rik; A630020C16Rik; TRFP; USP47
UniProt ID: (Human) Q96K76, (Mouse) Q8BY87
Entrez Gene ID: (Human) 55031, (Mouse) 74996
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support