Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Recommended positive controls: Raji.
Predicted reactivity: Mouse (100%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CD42a is a single-chain membrane glycoprotein that forms a noncovalent complex with CD42b. CD42b, also known as glycoprotein Ib a (GPIb a) is a membrane glycoprotein that is composed of a and b chains. The CD42b b chain is also designated CD42c, and is expressed on platelets and megakaryoctes. CD42a and CD42b are also present on platelets and mega-karyocytes, and the complex is a major component of the platelet surface. The complex acts as a receptor for von Willebrand's factor and as a von Willebrand's factor-dependent adhesion receptor.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Antigen CD42b-beta; CD42c; glycoprotein Ib (platelet), beta polypeptide; glycoprotein Ib platelet beta subunit; GP-Ib beta; GPIb-beta; GPIbB; nuclear localization signal deleted in velocardiofacial syndrome; Platelet glycoprotein Ib beta chain; platelet glycoprotein Ib beta chain precursor; platelet membrane glycoprotein Ib beta
Gene Aliases: BDPLT1; BS; CD42C; GP1BB; GPIBB; GPIbbeta
UniProt ID: (Human) P13224, (Mouse) P56400
Entrez Gene ID: (Human) 2812, (Mouse) 14724
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support