Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Recommended positive controls: 293T, HepG2.
Predicted reactivity: Mouse (92%), Rat (92%), Xenopus laevis (80%), Pig (94%), Rabbit (95%), Chicken (84%), Rhesus Monkey (98%), Bovine (95%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PHKG1 is a serine/threonine kinase with glycogenolytic regulatory functions. This protein is the catalytic member of a 16 subunit protein kinase complex which contains equimolar ratios of 4 subunit types. The complex is a crucial glycogenolytic regulatory enzyme. PHKG1 has two pseudogenes at chromosome 7q11.21 and one at chromosome 11p11.12.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PHK-gamma-M; PHK-gammaM; PHKgamma-M; PHKgammaM; Phosphorylase b kinase gamma catalytic chain, skeletal muscle/heart isoform; phosphorylase kinase; phosphorylase kinase gamma subunit 1; Phosphorylase kinase subunit gamma-1; phosphorylase kinase, gamma 1 (muscle); Serine/threonine-protein kinase PHKG1; unnamed protein product
Gene Aliases: PHKG; PHKG1
UniProt ID: (Human) Q16816
Entrez Gene ID: (Human) 5260
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support