Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Recommended positive controls: A549, HCT116, mouse brown adipose tissue.
Predicted reactivity: Mouse (89%), Rat (90%), Xenopus laevis (80%), Dog (97%), Chicken (86%), Sheep (96%), Bovine (96%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Adenylate kinase is an enzyme involved in regulating the adenine nucleotide composition within a cell by catalyzing the reversible transfer of phosphate group among adinine nucleotides. Three isozymes of adenylate kinase have been identified in vertebrates, adenylate isozyme 1 (AK1), 2 (AK2) and 3 (AK3). AK1 is found in the cytosol of skeletal muscle, brain and erythrocytes, whereas AK2 and AK3 are found in the mitochondria of other tissues including liver and heart. AK1 was identified because of its association with a rare genetic disorder causing nonspherocytic hemolytic anemia where a mutation in the AK1 gene was found to reduce the catalytic activity of the enzyme.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenylate kinase isoenzyme 1; Adenylate monophosphate kinase; AK 1; AK1; ATP-AMP transphosphorylase 1; ATP:AMP phosphotransferase; cytosolic adenylate kinase; epididymis secretory sperm binding protein; Myokinase; testis secretory sperm binding protein Li 58j; unnamed protein product
Gene Aliases: ADK; Adk1; Ak-1; AK1; B430205N08Rik; CNSHA3; HTL-S-58j
UniProt ID: (Human) P00568, (Mouse) Q9R0Y5
Entrez Gene ID: (Human) 203, (Mouse) 11636
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support