Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Recommended positive controls: mouse brain.
Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (94%), Chicken (94%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Sodium-hydrogen exchangers (NHEs), such as SLC9A8, are integral transmembrane proteins that exchange extracellular Na+ for intracellular H+. NHEs have multiple functions, including intracellular pH homeostasis, cell volume regulation, and electroneutral NaCl absorption in epithelia (Xu et al., 2008).
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686C03237; FLJ42500; MGC138418; Na(+)/H(+) exchanger 8; NHE-8; NHE-8 antibody; SLC9A8 antibody; Sodium/hydrogen exchanger 8; solute carrier family 9 (sodium/hydrogen exchanger), member 8; Solute carrier family 9 member 8; solute carrier family 9, subfamily A (NHE8, cation proton antiporter 8), member 8; unnamed protein product
Gene Aliases: 1200006P13Rik; 6430709P13Rik; AI182282; KIAA0939; NHE-8; NHE8; SLC9A8
UniProt ID: (Human) Q9Y2E8, (Mouse) Q8R4D1
Entrez Gene ID: (Human) 23315, (Mouse) 77031
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support