Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide sequence: VATFLYQVAE RVRYYREFHH QTALDERESH RLIFPAVTLC NINPLRRSRL
Sequence homology: Cow: 80%; Dog: 80%; Guinea Pig: 80%; Horse: 80%; Human: 100%; Mouse: 87%; Pig: 87%; Rabbit: 87%; Rat: 87%
ACCN3 encodes a member of the degenerin/epithelial sodium channel (DEG/ENaC) superfamily. The members of this family are amiloride-sensitive sodium channels that contain intracellular N and C termini, two hydrophobic transmembrane regions, and a large extracellular loop, which has many cysteine residues with conserved spacing. ACCN3 encodes a member that is an acid sensor and may play an important role in the detection of lasting pH changes. In addition, a heteromeric association between this member and ACCN1 has been observed as proton-gated channels sensitive to gadolinium.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acid sensing (proton gated) ion channel 3; Acid-sensing ion channel 3; Amiloride-sensitive cation channel 3; amiloride-sensitive cation channel 3, testis; ASIC3; ASIC3b; ASIC3c; hASIC3; hTNaC1; modulatory subunit of ASIC2a; Neuronal amiloride-sensitive cation channel 3; proton-gated cation channel subunit; Testis sodium channel 1; unnamed protein product
Gene Aliases: ACCN3; ASIC3; DRASIC; SLNAC1; TNAC1
UniProt ID: (Human) Q9UHC3
Entrez Gene ID: (Human) 9311
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support