Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide sequence: MLEGQLEAGE PKEGTHPEDP CPGAGAAMEK TPAAAEVPRE DSNAGEMPSL
Sequence homology: Human: 100%
The exact function of TCP10 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene Aliases: TCP10; TCP10A
Entrez Gene ID: (Human) 6953
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support