Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide sequence: GVMMLQLLEK VIELENWTEG KGLIVRGAKN TFSSGSDLNA VKSLGTPEDG
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 92%; Rabbit: 93%; Rat: 100%
ECHDC1 belongs to the enoyl-CoA hydratase/isomerase family. The exact function of ECHDC1 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: enoyl CoA hydratase domain containing 1; enoyl Coenzyme A hydratase domain containing 1; Enoyl-CoA hydratase domain-containing protein 1; epididymis secretory protein Li 76; Ethylmalonyl-CoA decarboxylase; Methylmalonyl-CoA decarboxylase; MMCD; unnamed protein product
Gene Aliases: dJ351K20.2; ECHDC1; HEL-S-76; MMCD
UniProt ID: (Human) Q9NTX5
Entrez Gene ID: (Human) 55862
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support