Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide sequence: NANNFYTWEA CDDACWRIEK VPKVCRLQVS VDDQCEGSTE KYFFNLSSMT
Sequence homology: Human: 100%
TFPI2 may play a role in the regulation of plasmin-mediated matrix remodeling. It inhibits trypsin, plasmin, factor VIIa/tissue factor and weakly factor Xa. TFPI2 has no effect on thrombin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: asTFPI2; Placental protein 5; PP5; PP5FLJ21164; REF-1; retinal pigment epithelium cell factor 1; TFPI-2; TFPI-2REF1; Tissue factor pathway inhibitor 2; unnamed protein product
Gene Aliases: PP5; REF1; TFPI-2; TFPI2
UniProt ID: (Human) P48307
Entrez Gene ID: (Human) 7980
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support