Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide sequence: GAQALALRRM QDEPYQALVA ELHRRALVEY VRPLLRGRLR CSSARTRSRV
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Human: 100%; Mouse: 93%; Rat: 100%
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Exocyst complex component 3-like protein 2; HBV X-transactivated gene 7 protein; HBV XAg-transactivated protein 7; protein 7 transactivated by hepatitis B virus X antigen (HBxAg); unnamed protein product
Gene Aliases: BMRS; EXOC3L2; XTP7
UniProt ID: (Human) Q2M3D2
Entrez Gene ID: (Human) 90332
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support