Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The antibody detects endogenous levels of total DOPEY1 protein.
May be involved in protein traffic between late Golgi and early endosomes. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dopey family member 1; homolog of yeast DOP1; Protein DOP1A; protein dopey-1; unnamed protein product
Gene Aliases: B130005I07Rik; BC035275; C130028L17; D9Ertd809e; dJ202D23.2; DOP1; DOP1A; DOPEY1; KIAA1117
UniProt ID: (Human) Q5JWR5
Entrez Gene ID: (Human) 23033, (Mouse) 320615
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support