Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: PEVCTCVPGG LASCSALSLP AVPPGLSLRL RALLLDHNRV RA
Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 67%.
The LRRC26 protein was initially identified as a cytokeratin-associated protein whose expression was elevated in multiple cancer cell lines and cancer specimens as well in normal prostate and salivary gland tissues. LRRC26 was later found to negatively regulate the NF-kappaB activation and suppresses tumor growth and metastasis. LRRC26 can also act to selectively alter the efficacy of large conductance calcium-activated K (BK) channels.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BK channel auxiliary gamma subunit LRRC26; BK channel auxilliary gamma subunit LRRC26; CAPC; cytokeratin associated protein; cytokeratin-associated protein in cancer
Gene Aliases: bA350O14.10; CAPC
UniProt ID: (Human) Q2I0M4
Entrez Gene ID: (Human) 389816
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support