Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: EDKRHAFRNGG GPAYQLREWK EPASYGRQDA TVWIPSPSRQ PVYYDELDAA SSSLRRLSLQ PRSHSVPRSP SQGSYSRARI YSPVRSPSAR F
Highest antigen sequence identity to the following orthologs - mouse 91%, rat 29%.
PLEKHA6 (Pleckstrin Homology Domain Containing A6) is a Protein Coding gene. Among its related pathways arePI MetabolismandGlycerophospholipid biosynthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PEPP-3; PH domain-containing family A member 6; Phosphoinositol 3-phosphate-binding protein 3; Pleckstrin homology domain-containing family A member 6; unnamed protein product
Gene Aliases: KIAA0969; PEPP-3; PEPP3; PLEKHA6
UniProt ID: (Human) Q9Y2H5
Entrez Gene ID: (Human) 22874
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support