Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
LCN6 is a member of the lipocalin family of hydrophobic ligand binding proteins and exhibit conserved beta strand basket like structures. LCN6 may play a role in male fertility.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymal-specific lipocalin ELP16; epididymal-specific lipocalin LCN6; Epididymal-specific lipocalin-6; epididymis secretory sperm binding protein; hLcn5; lipocalin 5; Lipocalin-5; MCG18768
Gene Aliases: 9230101D24Rik; hLcn5; LCN5; LCN6; UNQ643; UNQ643/PRO1273
UniProt ID: (Human) P62502
Entrez Gene ID: (Human) 158062, (Rat) 414138, (Mouse) 620709
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support