Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
FKBP6 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. FKBP6 is a cis-trans prolyl isomerase that binds to the immunosuppressants FK506 and rapamycin.This gene encodes a member of the interferon regulatory transcription factor (IRF) family. Family members share a highly-conserved N-terminal helix-turn-helix DNA-binding domain and a less conserved C-terminal protein-binding domain. Mutations in this gene can cause van der Woude syndrome and popliteal pterygium syndrome. This protein is involved in palate formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 36 kDa FK506 binding protein; 36 kDa FK506-binding protein; 36 kDa FKBP; 36kD FKBP6; FK506 binding protein 6, 36kDa; FK506-binding protein 6; FK506-binding protein 6 (36kD); FKBP prolyl isomerase 6; FKBP-6; Immunophilin FKBP36; Inactive peptidyl-prolyl cis-trans isomerase FKBP6; Inactive PPIase FKBP6; MGC87179; peptidyl-prolyl cis-trans isomerase FKBP6; PPIase FKBP6; rotamase; unnamed protein product
Gene Aliases: 1700008G22Rik; 36kDa; As; AU017274; D5Ertd724e; FKBP-36; FKBP-6; FKBP36; FKBP6; PPiase; SPGF77
UniProt ID: (Human) O75344, (Mouse) Q91XW8, (Rat) D3ZQF4
Entrez Gene ID: (Human) 8468, (Mouse) 94244, (Rat) 288597
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support