Search
Search
FIGURE: 1 / 7
Positive Control: 293T, mouse brain, rat brain
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NEK1 is a serine/threonine protein kinase, related to the NIMA cell cycle regulator, and is highly expressed in meiotic germ cells. The kinase may function early in the DNA damage response pathway. NEK1 may also participate in polycystic kidney disease and neural cell development. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686D06121; DKFZp686K12169; kidney, anemia and testis; MGC138800; Nek1 serine/threonine- and tyrosine-specific protein kinase; Never in mitosis A-related kinase 1; NIMA (never in mitosis gene a)-related kinase 1; NIMA-related kinase 1; NimA-related protein kinase 1; NY REN55; NY-REN-55 antigen; protein-serine/threonine kinase; Renal carcinoma antigen NY-REN-55; Serine/threonine-protein kinase Nek1; unnamed protein product
Gene Aliases: ALS24; D8Ertd790e; kat; KIAA1901; NEK1; NY-REN-55; OFD2; SRPS2; SRPS2A; SRTD6
UniProt ID: (Human) Q96PY6, (Mouse) P51954
Entrez Gene ID: (Human) 4750, (Mouse) 18004
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support