Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 14
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
Positive Control - WB: human A549 whole cell, human MOLT4 whole cell, human U87 whole cell, human U251 whole cell, human 293T whole cell, human Hela whole cell, human T47D whole cell, human HepG2 whole cell, rat pancreas tissue, mouse NIH/3T3 whole cell. IHC: human colonic adenocarcinoma tissue, human hepatocellular carcinoma tissue, human laryngeal squamous cell carcinoma tissue, human placenta tissue, human colon cancer tissue, mouse testis tissue, rat testis tissue. ICC/IF: A594 cell. Flow: HepG2 cell.
ATP citrate lyase is the primary enzyme responsible for the synthesis of cytosolic acetyl-CoA in many tissues. The enzyme is a tetramer (relative molecular weight approximately 440,000) of apparently identical subunits. It catalyzes the formation of acetyl-CoA and oxaloacetate from citrate and CoA with a concomitant hydrolysis of ATP to ADP and phosphate. The product, acetyl-CoA, serves several important biosynthetic pathways, including lipogenesis and cholesterogenesis. In nervous tissue, ATP citrate-lyase may be involved in the biosynthesis of acetylcholine. Two transcript variants encoding distinct isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACL; ATP-citrate (pro-S-)-lyase; ATP-citrate synthase; Citrate cleavage enzyme; unnamed protein product
Gene Aliases: A730098H14Rik; ACL; ACLY; ATPCL; AW538652; CLATP
UniProt ID: (Human) P53396, (Mouse) Q91V92, (Rat) P16638
Entrez Gene ID: (Human) 47, (Mouse) 104112, (Rat) 24159
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support