Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: IRSRRSSQRA VLLVGLCDSG KTLLFVRLLT GLYRDTQTSI TDSCAVYRVN NNRGNSLTLI DLPGHESLRL QFLERFKSSA RAIVFVVDSA AFQREVKDVA EFLYQVLIDS MGLKNTPSFL IACNKQDIAM AKSAKLIQQQ LEKELNTLRV TRSAAPSTLD SSSTAPAQLG KKGKEFEFSQ LPLKVEFLEC SAKGGRGDVG SADIQDLEKW LAKIA; Positive Samples: MCF7, HT-29, LO2, , B cells, Mouse liver, Mouse pancreas, Mouse small intestine, Rat liver; Cellular Location: Endoplasmic reticulum membrane, Single-pass membrane protein
SRPRB has similarity to mouse protein which is a subunit of the signal recognition particle receptor (SR). This subunit is a transmembrane GTPase belonging to the GTPase superfamily. It anchors alpha subunit, a peripheral membrane GTPase, to the ER membrane. SR is required for the cotranslational targeting of both secretory and membrane proteins to the ER membrane. The protein encoded by this gene has similarity to mouse protein which is a subunit of the signal recognition particle receptor (SR). This subunit is a transmembrane GTPase belonging to the GTPase superfamily. It anchors alpha subunit, a peripheral membrane GTPase, to the ER membrane. SR is required for the cotranslational targeting of both secretory and membrane proteins to the ER membrane. The protein encoded by this gene has similarity to mouse protein which is a subunit of the signal recognition particle receptor (SR). This subunit is a transmembrane GTPase belonging to the GTPase superfamily. It anchors alpha subunit, a peripheral membrane GTPase, to the ER membrane. SR is required for the cotranslational targeting of both secretory and membrane proteins to the ER membrane.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein APMCF1; Signal recognition particle receptor subunit beta; signal recognition particle receptor, B subunit; signal recognition particle receptor, beta subunit; SR-beta; SRP receptor beta subunit; SRPR beta; unnamed protein product
Gene Aliases: AA409543; Ab2-417; APMCF1; Ba1-667; Cc1-8; PSEC0230; SR-beta; SRPRB
UniProt ID: (Human) Q9Y5M8, (Rat) Q4FZX7, (Mouse) P47758
Entrez Gene ID: (Human) 58477, (Rat) 300965, (Mouse) 20818
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support