Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: RCLLNTSNHS GVEAFIIQNI KNQIDMSLKR TRNNKWFTGP QLISLLDLVL FLPEGAETDL LQNSDRIMAS LNLLRYLVIK DNENDNQTGL WTELGNIENN FLKPLHIGLN MSKAHYEAEI KNSQEAQKSK DLCSITVSGE EIPNMPPEMQ LKVLHSALFT FDLIESVLAR VEELIEIKTK STSEENIGIK; Positive Samples: A-549, U-87MG, HepG2, Mouse testis
This gene encodes a phosphorylated protein that is a member of a Skp1-Cullin-F-box-like complex. The protein is essential for normal development of the vasculature and mutations in this gene have been associated with glomuvenous malformations, also called glomangiomas. Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only one has been determined.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FAP; FK506-binding protein-associated protein; FKBP-associated protein; Glomulin; unnamed protein product
Gene Aliases: 9330160J16Rik; AW227515; FAP; FAP48; FAP68; FKBPAP; GLML; GLMN; GVM; VMGLOM
UniProt ID: (Human) Q92990, (Mouse) Q8BZM1
Entrez Gene ID: (Human) 11146, (Mouse) 170823
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support