Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: GLTYEKLNRL DEALDCFLKL HAILRNSAEV LYQIANIYEL MENPSQAIEW LMQVVSVIPT DPQVLSKLGE LYDREGDKSQ AFQYYYESYR YFPCNIEVIE WLGAYYIDTQ FWEKAIQYFE RASLIQPTQV KWQLMVASCF RRSGNYQKAL DTYKDTHRKF PENVECLRFL VRLCTDLGLK DAQEYARKLK RLEKMKEIRE QRIKSGRDGS GGSRGKREGS ASGDSGQNYS ASSKGERLSA RLRALPGTNE PYESSSNKEI DASYVDPLGP QIERPKTAAK KRIDEDDFAD EELGDDLLPE; Positive Samples: HepG2, BxPC-3, Mouse brain, Mouse eye; Cellular Location: Cytoplasm, centriole, centrosome, cilium basal body, cytoskeleton, microtubule organizing center
This gene encodes a member of the tetratrico peptide repeat family. Mutations of a similar gene in mouse can cause polycystic kidney disease. Two transcript variants encoding distinct isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HGNC:20606; intraflagellar transport 88 homolog; Intraflagellar transport protein 88 homolog; MGC26259; mutations in the mouse Tg737 gene cause polycystic kidney disease; polaris homolog; probe hTg737 (polycystic kidney disease, autosomal recessive); Recessive polycystic kidney disease protein Tg737; Recessive polycystic kidney disease protein Tg737 homolog; RP11-172H24.2; testicular tissue protein Li 93; tetratricopeptide repeat domain 10; Tetratricopeptide repeat protein 10; Tg737 protein; TgN(Imorpk)737Rpw; TPR repeat protein 10; transgene insert site 737, insertional mutation, polycystic kidney disease; unnamed protein product
Gene Aliases: AW552028; D13S1056E; DAF19; flexo; fxo; hTg737; IFT88; orpk; polaris; TG737; Tg737Rpw; TgN737Rpw; TTC10
UniProt ID: (Human) Q13099, (Mouse) Q61371
Entrez Gene ID: (Human) 8100, (Mouse) 21821
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support