Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: MWATCCNWFC LDGQPEEVPP PQGARMQAYS NPGYSSFPSP TGLEPSCKSC GAHFANTARK QTCLDCKKNF CMTCSSQVGN GPRLCLLCQR FRATAFQREE LMKMKVKDLR DYLSLHDIST EMCREKEELV LLVLGQQPVI SQEDRTRAST LSPDFPEQQA FLTQPHSSMV PPTSPNLPSS SAQATSVPPA; Positive Samples: U-251MG; Cellular Location: Cell membrane, Cytoplasm, Peripheral membrane protein, Recycling endosome membrane, cytosol
Has E3 ubiquitin protein ligase activity. Regulates the levels of CASP8 and CASP10 by targeting them for proteasomal degradation. Has anti-apoptotic activity. May bind phosphatidylinositol phosphates.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CARP-2; caspase 8 and 10 associated RING protein-2; Caspase regulator CARP2; Caspases-8 and -10-associated RING finger protein 2; E3 ubiquitin-protein ligase rififylin; Fring; FYVE-RING finger protein SAKURA; rififylin; ring finger and FYVE like domain containing protein; ring finger and FYVE-like domain containing 1; RING finger and FYVE-like domain-containing protein 1; RING finger protein 189; RING finger protein 34-like; RING finger protein SAKURA; RING-type E3 ubiquitin transferase rififylin; unnamed protein product
Gene Aliases: 1700051E09Rik; 4930516L10Rik; BG080975; CARP-2; CARP2; FRING; RFFL; RIFIFYLIN; RNF189; RNF34L
UniProt ID: (Human) Q8WZ73, (Mouse) Q6ZQM0, (Rat) Q8CIN9
Entrez Gene ID: (Human) 117584, (Mouse) 67338, (Rat) 282844
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support